N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 95%

Supplier: Ambeed
Danger

Synonyms: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate

A140481-10G A140481-1G A140481-25G A140481-500G A140481-5G
76996-846EA 15 USD
76996-846 76996-848 76996-850 76996-852 76996-854
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 95%
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₆BF₄N₃O₃
MDL Number: MFCD00077875
CAS Number: 105832-38-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR