Di-tert-butyl (azanediylbis(propane-3,1-diyl))dicarbamate 97%

Supplier: Ambeed
Danger

A140455-100MG A140455-250MG A140455-5G A140455-1G
77010-334EA 55.17 USD
77010-334 77010-336 77566-830 77592-666
Di-tert-butyl (azanediylbis(propane-3,1-diyl))dicarbamate 97%
Di-tert-butyl (azanediylbis(propane-3,1-diyl))dicarbamate

Precaución: For research use only.

Formula: C₁₆H₃₃N₃O₄
MW: 331.45 g/mol
MDL Number: MFCD11226826
CAS Number: 82409-02-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR