3-(Morpholinomethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-(Morpholinomethyl)phenyl)boronic acid

A140338-1G A140338-250MG
77010-330EA 28 USD
77010-330 77010-332
3-(Morpholinomethyl)phenylboronic acid 97%
3-(Morpholinomethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MDL Number: MFCD08060623
CAS Number: 397843-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR