2-Fluoro-3-nitro-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methyl-3-nitropyridine

A140227-1G A140227-5G
76996-840EA 38 USD
76996-840 76996-842
2-Fluoro-3-nitro-4-picoline 98%
2-Fluoro-3-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD04112526
CAS Number: 19346-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR