Methyl 3-(1H-indol-3-yl)propanoate 95%
Supplier: Ambeed
|
Warning
|
A140204-100MG
A140204-1G
A140204-250MG
A140204-5G
76984-144EA
6.37
USD
76984-144
76984-146
76984-148
77566-814
Methyl 3-(1H-indol-3-yl)propanoate 95%
Methyl 3-(1H-indol-3-yl)propanoate
Precaución: For research use only.
Formula:
C₁₂H₁₃NO₂ MW: 203.24 g/mol |
MDL Number:
MFCD00022778 CAS Number: 5548-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...