2-Bromo-6-(difluoromethoxy)pyridine 97%

Supplier: Ambeed
Warning

A140189-250MG A140189-5G A140189-1G
76984-142EA 37.14 USD
76984-142 77566-808 77683-224
2-Bromo-6-(difluoromethoxy)pyridine 97%
2-Bromo-6-(difluoromethoxy)pyridine

Precaución: For research use only.

Formula: C₆H₄BrF₂NO
MW: 224.00 g/mol
MDL Number: MFCD13185843
CAS Number: 1214345-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR