3-Iodo-5-(trifluoromethyl)benzoic acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A140147-1G A140147-250MG A140147-5G A140147-25G
77010-324EA 35 USD
77010-324 77010-326 77010-328 77683-223
3-Iodo-5-(trifluoromethyl)benzoic acid 95%, Ambeed.Inc
3-Iodo-5-(trifluoromethyl)benzoic acid

Precaución: For research use only.

Formula: C₈H₄F₃IO₂
MDL Number: MFCD08448293
CAS Number: 28186-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR