4-Methoxyphenethyl bromide 98%

Supplier: Ambeed
Warning

A140110-1G A140110-25G A140110-5G A140110-250MG
77027-466EA 22 USD
77027-466 77027-468 77549-704 77671-000
4-Methoxyphenethyl bromide 98%
4-Methoxyphenethyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD07369786
CAS Number: 14425-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR