5-Bromo-2-chlorobenzothiazole 98%

Supplier: Ambeed
Warning

A140063-100MG A140063-250MG A140063-5G A140063-1G
77027-458EA 13 USD
77027-458 77027-460 77667-024 77871-674
5-Bromo-2-chlorobenzothiazole 98%
5-Bromo-2-chlorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MDL Number: MFCD09749227
CAS Number: 824403-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR