3,5-Di-tert-butyl-2-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Di-tert-butylsalicylaldehyde
A140025-100G
A140025-25G
A140025-5G
77027-448EA
20.16
USD
77027-448
77027-450
77027-452
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 97%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₂₂O₂ MW: 234.34 g/mol |
MDL Number:
MFCD00191998 CAS Number: 37942-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...