N2-Boc-3-methoxy-1,2-benzenediamine 96%

Supplier: Ambeed
Warning

A139858-100MG A139858-1G A139858-250MG
77027-434EA 31 USD
77027-434 77027-436 77027-438
N2-Boc-3-methoxy-1,2-benzenediamine 96%
N2-Boc-3-methoxy-1,2-benzenediamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₃
MW: 238.283 g/mol
MDL Number: MFCD09701243
CAS Number: 954238-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR