Methyl isoquinoline-1-carboxylate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A139726-1G A139726-5G A139726-25G
76984-134EA 35 USD
76984-134 77683-218 77889-946
Methyl isoquinoline-1-carboxylate 95%, Ambeed.Inc
Methyl isoquinoline-1-carboxylate

Precaución: For research use only.

Formula: C₁₁H₉NO₂
MDL Number: MFCD11506138
CAS Number: 27104-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR