6-Chloro-5-isopropylpyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A139657-100MG A139657-1G A139657-250MG
77010-316EA 63.66 USD
77010-316 77010-318 77010-320
6-Chloro-5-isopropylpyrimidin-4-amine 97%
6-Chloro-5-isopropylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₀ClN₃
MW: 171.63 g/mol
MDL Number: MFCD18874248
CAS Number: 852061-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR