Fmoc-D-4-bromophenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-bromo-D-Phe-OH, 4-Bromo-N-Fmoc-D-phenylalanine, Fmoc-D-Phe(4-Br)-OH

A139592-10G A139592-1G A139592-250MG A139592-25G A139592-5G
77027-420EA 57 USD
77027-420 77027-422 77027-424 77027-426 77027-428
Fmoc-D-4-bromophenylalanine 97%
Fmoc-D-4-bromophenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀BrNO₄
MW: 466.331 g/mol
MDL Number: MFCD00273459
CAS Number: 198545-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR