(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-bromopyridin-3-yl)carbamate

A139523-100MG A139523-250MG A139523-1G
77010-304EA 49.87 USD
77010-304 77010-306 77592-648
(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester 98%
(2-Bromopyridin-3-yl)carbamic acid-tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.14 g/mol
MDL Number: MFCD07367911
CAS Number: 116026-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR