Methyl 6-chloro-2-fluoronicotinate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-fluoro-nicotinic acid methyl ester

A139488-250MG A139488-5G A139488-1G
76984-130EA 34 USD
76984-130 77683-216 77885-179
Methyl 6-chloro-2-fluoronicotinate 95%, Ambeed.Inc
Methyl 6-chloro-2-fluoronicotinate

Precaución: For research use only.

Formula: C₇H₅ClFNO₂
MDL Number: MFCD16658093
CAS Number: 1093880-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR