(3S,4S)-3-Amino-4-hydroxytetrahydropyran 98%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-3-Aminotetrahydro-2H-pyran-4-ol

A139365-100MG A139365-250MG
77010-300EA 57.29 USD
77010-300 77010-302
(3S,4S)-3-Amino-4-hydroxytetrahydropyran 98%
(3S,4S)-3-Amino-4-hydroxytetrahydropyran

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD19215826
CAS Number: 1240390-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR