3,4-Dibromophenylboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A139190-100MG A139190-1G A139190-250MG
77010-294EA 36.08 USD
77010-294 77010-296 77010-298
3,4-Dibromophenylboronic acid pinacol ester 96%
3,4-Dibromophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BBr₂O₂
MW: 361.87 g/mol
MDL Number: MFCD12026084
CAS Number: 1075719-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR