2-(4-Methoxyphenyl)-1,3,4-oxadiazole 97%

Supplier: Ambeed
Warning

A139158-100MG A139158-1G
77027-414EA 46.68 USD
77027-414 77027-416
2-(4-Methoxyphenyl)-1,3,4-oxadiazole 97%
2-(4-Methoxyphenyl)-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
MDL Number: MFCD00491615
CAS Number: 829-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR