4-Amino-2-chloropyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-chloro-5-pyrimidinecarbonitrile

A139154-1G A139154-250MG A139154-5G
76984-122EA 19.1 USD
76984-122 76984-124 77611-262
4-Amino-2-chloropyrimidine-5-carbonitrile 97%
4-Amino-2-chloropyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MW: 154.56 g/mol
MDL Number: MFCD00052343
CAS Number: 94741-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR