2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%

Supplier: Ambeed
Warning

Synonyms: Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 2,2-Dimethyltetrahydropyran-4-one

A139082-100G A139082-1G A139082-250MG A139082-25G A139082-5G
77010-284EA 758.55 USD
77010-284 77010-286 77010-288 77010-290 77010-292
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD01549337
CAS Number: 1194-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR