2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2-Di-t-butylphosphino-2'-(N,N-dimethylamino))-1,1'-biphenyl, tBuDavePhos, 2'-(Di-tert-butylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine

A139069-10G A139069-1G A139069-250MG A139069-5G
76996-802EA 257.8 USD
76996-802 76996-804 76996-806 76996-808
2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl 98%
2-(Di-tert-butylphosphino)-2'-(N,N-dimethylamino)biphenyl

Precaución: for Research Use Only

Formula: C₂₂H₃₂NP
MW: 341.47 g/mol
MDL Number: MFCD03426986
CAS Number: 224311-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR