1,4-Bis(7-chloroquinolin-4-yl)piperazine 95%

Supplier: Ambeed
Warning

A139012-100MG A139012-1G A139012-250MG
76984-104EA 31.83 USD
76984-104 76984-106 76984-108
1,4-Bis(7-chloroquinolin-4-yl)piperazine 95%
1,4-Bis(7-chloroquinolin-4-yl)piperazine

Precaución: for Research Use Only

Formula: C₂₂H₁₈Cl₂N₄
MW: 409.31 g/mol
MDL Number: MFCD01236858
CAS Number: 31502-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR