5-Phenyl-1H-imidazole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A138914-100MG A138914-1G A138914-250MG
76984-098EA 168.69 USD
76984-098 76984-100 76984-102
5-Phenyl-1H-imidazole-2-carbaldehyde 95%
5-Phenyl-1H-imidazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
MDL Number: MFCD10696709
CAS Number: 56248-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR