N-Methylhex-5-en-1-amine hydrochloride 97%

Supplier: Ambeed
Warning

A138752-1G A138752-250MG
77010-260EA 152.77 USD
77010-260 77010-262
N-Methylhex-5-en-1-amine hydrochloride 97%
N-Methylhex-5-en-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClN
MW: 149.66 g/mol
MDL Number: MFCD19982659
CAS Number: 862175-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR