5-Nitro-2,3-pyridinediamine 95%

Supplier: Ambeed
Warning

A138711-100MG A138711-250MG A138711-5G A138711-1G
77027-404EA 97 USD
77027-404 77027-406 77027-408 77666-996
5-Nitro-2,3-pyridinediamine 95%
5-Nitro-2,3-pyridinediamine

Precaución: for Research Use Only

Formula: C₅H₆N₄O₂
MDL Number: MFCD10697697
CAS Number: 3537-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR