2-Chloro-4-ethynylpyridine 98%

Supplier: Ambeed
Warning

A138674-100MG A138674-10G A138674-1G A138674-5G
77027-396EA 26.53 USD
77027-396 77027-398 77027-400 77027-402
2-Chloro-4-ethynylpyridine 98%
2-Chloro-4-ethynylpyridine

Precaución: for Research Use Only

Formula: C₇H₄ClN
MW: 137.5664 g/mol
MDL Number: MFCD13189742
CAS Number: 945717-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR