3-Bromo-2-isopropoxy-5-(trifluoromethyl)pyridine 95+%

Supplier: Ambeed
Warning

A138504-1G A138504-5G A138504-25G
76984-088EA 14 USD
76984-088 76984-090 77889-938
3-Bromo-2-isopropoxy-5-(trifluoromethyl)pyridine 95+%
3-Bromo-2-isopropoxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₉BrF₃NO
MW: 284.07 g/mol
MDL Number: MFCD26743634
CAS Number: 216766-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR