4-Methyl-6-nitroindoline 97%

Supplier: Ambeed
Warning

A138459-100MG A138459-1G A138459-250MG
76984-080EA 67.9 USD
76984-080 76984-082 76984-084
4-Methyl-6-nitroindoline 97%
4-Methyl-6-nitroindoline

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
MDL Number: MFCD26743708
CAS Number: 1588441-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR