4-Bromo-3,5-dichloroaniline 95%

Supplier: Ambeed
Warning

A138440-1G A138440-5G
76984-078EA 48.81 USD
76984-078 77683-214
4-Bromo-3,5-dichloroaniline 95%
4-Bromo-3,5-dichloroaniline

Precaución: For research use only.

Formula: C₆H₄BrCl₂N
MW: 240.9100 g/mol
MDL Number: MFCD09878195
CAS Number: 1940-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR