4-Methoxy-4-methyl-piperidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-4-methylpiperidine hydrochloride

A138414-100MG A138414-1G A138414-250MG A138414-5G
77027-378EA 51.99 USD
77027-378 77027-380 77027-382 77027-384
4-Methoxy-4-methyl-piperidine hydrochloride 98%
4-Methoxy-4-methyl-piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO·ClH
MW: 165.661 g/mol
MDL Number: MFCD12755739
CAS Number: 3970-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR