3,4-Difluoro-5-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A138402-10G A138402-1G A138402-250MG A138402-5G
77010-238EA 500 USD
77010-238 77010-240 77010-242 77010-244
3,4-Difluoro-5-hydroxybenzaldehyde 97%
3,4-Difluoro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MDL Number: MFCD04972451
CAS Number: 887584-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR