Ethyl 2-(3-nitrophenyl)acetate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A138253-250MG A138253-5G A138253-1G
76984-070EA 6 USD
76984-070 77592-628 77683-213
Ethyl 2-(3-nitrophenyl)acetate 95%, Ambeed.Inc
Ethyl 2-(3-nitrophenyl)acetate

Precaución: For research use only.

Formula: C₁₀H₁₁NO₄
MW: 209.20 g/mol
MDL Number: MFCD09953138
CAS Number: 14318-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR