H-N-Me-D-Val-OH.HCl 97%
Supplier: Ambeed
|
Warning
|
A138157-10G
A138157-1G
A138157-250MG
A138157-5G
77010-224EA
136
USD
77010-224
77010-226
77010-228
77010-230
H-N-Me-D-Val-OH.HCl 97%
H-N-Me-D-Val-OH.HCl
Precaución: for Research Use Only
Formula:
C₆H₁₄ClNO₂ MW: 167.63 g/mol |
MDL Number:
MFCD00270621 CAS Number: 210830-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...