6-Aminopurine-8(9H)-thione 98%

Supplier: Ambeed
Warning

A137922-100MG A137922-250MG
76996-766EA 58.35 USD
76996-766 76996-768
6-Aminopurine-8(9H)-thione 98%
6-Aminopurine-8(9H)-thione

Precaución: for Research Use Only

Formula: C₅H₅N₅S
MW: 167.19 g/mol
MDL Number: MFCD00127786
CAS Number: 7390-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR