Propionyl-L-carnitine hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: Propionyl-L-carnitine HCl, (R)-3-(Propionyloxy)-4-(trimethylammonio)butanoate hydrochloride, PLC

A137851-100MG A137851-1G A137851-5G
76996-762EA 5.31 USD
76996-762 76996-764 77670-972
Propionyl-L-carnitine hydrochloride 96%
Propionyl-L-carnitine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄·HCl
MW: 253.8 g/mol
MDL Number: MFCD00274137
CAS Number: 119793-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR