Isoindolin-4-ol hydrochloride 97%

Supplier: Ambeed
Warning

A137845-100MG A137845-1G A137845-250MG A137845-5G
76984-056EA 21 USD
76984-056 76984-058 76984-060 76984-062
Isoindolin-4-ol hydrochloride 97%
Isoindolin-4-ol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
MDL Number: MFCD10700136
CAS Number: 72695-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR