2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethyl benzoate 98%

Supplier: Ambeed
Warning

A137827-1000G A137827-100G A137827-1MG A137827-25G A137827-500G
76996-752EA 198 USD
76996-752 76996-754 76996-756 76996-758 76996-760
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethyl benzoate 98%
2-(2-Methyl-5-nitro-1H-imidazol-1-yl)ethyl benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃O₄
MDL Number: MFCD00693654
CAS Number: 13182-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR