tert-Butyl 4-cyano-4-(4-fluorobenzyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A137825-100MG A137825-250MG
77010-202EA 19.1 USD
77010-202 77010-204
tert-Butyl 4-cyano-4-(4-fluorobenzyl)piperidine-1-carboxylate 97%
tert-Butyl 4-cyano-4-(4-fluorobenzyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₃FN₂O₂
MW: 318.39 g/mol
MDL Number: MFCD11849799
CAS Number: 894769-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR