(R)-Ethyl-3-piperidinecarboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A137813-1G
A137813-5G
A137813-10G
A137813-25G
76996-748EA
5
USD
76996-748
76996-750
77881-084
77881-085
(R)-Ethyl-3-piperidinecarboxylate hydrochloride 97%
(R)-Ethyl 3-piperidinecarboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₆ClNO₂ |
MDL Number:
MFCD05864623 CAS Number: 37675-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...