4,6-Dichloro-3-nitropyridin-2-amine 97%

Supplier: Ambeed
Warning

A137681-250MG A137681-1G A137681-5G A137681-100MG
76984-054EA 21 USD
76984-054 77566-762 77566-764 77889-928
4,6-Dichloro-3-nitropyridin-2-amine 97%
4,6-Dichloro-3-nitropyridin-2-amine

Precaución: For research use only.

Formula: C₅H₃Cl₂N₃O₂
MDL Number: MFCD26383593
CAS Number: 37660-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR