4-Bromo-3-ethoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A137598-10G A137598-1G A137598-250MG A137598-5G
77010-194EA 143.23 USD
77010-194 77010-196 77010-198 77010-200
4-Bromo-3-ethoxybenzaldehyde 97%
4-Bromo-3-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
MDL Number: MFCD22383779
CAS Number: 1353962-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR