5,6-Dichloro-3-nitropyridin-2-amine 95%

Supplier: Ambeed
Warning

A137551-100MG A137551-1G A137551-250MG
76984-042EA 35.01 USD
76984-042 76984-044 76984-046
5,6-Dichloro-3-nitropyridin-2-amine 95%
5,6-Dichloro-3-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N₃O₂
MW: 208.00 g/mol
MDL Number: MFCD22987594
CAS Number: 203794-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR