3-Chloro-5-hydroxy-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A137404-1G A137404-250MG
77010-180EA 363.89 USD
77010-180 77010-182
3-Chloro-5-hydroxy-4-methoxybenzaldehyde 95%
3-Chloro-5-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD01191266
CAS Number: 54246-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR