5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Oxoisoindoline-5-boronic acid, pinacol ester
A137391-100MG
A137391-1G
A137391-250MG
A137391-5G
76996-708EA
41.38
USD
76996-708
76996-710
76996-712
76996-714
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₃ MW: 259.11 g/mol |
MDL Number:
MFCD11040393 CAS Number: 376584-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...