1-(3-Aminophenyl)-3-methyl-2-imidazolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Aminophenyl)-3-methylimidazolidin-2-one

A137384-100MG A137384-1G A137384-250MG
77027-348EA 43.5 USD
77027-348 77027-350 77027-352
1-(3-Aminophenyl)-3-methyl-2-imidazolidinone 98%
1-(3-Aminophenyl)-3-methyl-2-imidazolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃O
MW: 191.2300 g/mol
MDL Number: MFCD05864562
CAS Number: 517918-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR