2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole 95%

Supplier: Ambeed
Warning

A137317-250MG A137317-1G
77027-346EA 9 USD
77027-346 77666-978
2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole 95%
2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole

Precaución: for Research Use Only

Formula: C₉H₇F₃N₂O
MDL Number: MFCD12755183
CAS Number: 6758-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR