Ethyl pyrazine-2-carboxylate 98%

Supplier: Ambeed
Warning

A137195-10G A137195-1G A137195-250MG A137195-25G A137195-5G
77027-336EA 51 USD
77027-336 77027-338 77027-340 77027-342 77027-344
Ethyl pyrazine-2-carboxylate 98%
Ethyl pyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00792887
CAS Number: 6924-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR