3-Methyl-3-phenylbutanal 97%

Supplier: Ambeed
Warning

A136995-100MG A136995-1G
77010-170EA 110 USD
77010-170 77010-172
3-Methyl-3-phenylbutanal 97%
3-Methyl-3-phenylbutanal

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MDL Number: MFCD00156879
CAS Number: 6325-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR