2-Amino-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A136969-250MG A136969-100MG A136969-1G
77027-334EA 47 USD
77027-334 77599-990 77670-964
2-Amino-7H-pyrrolo[2,3-d]pyrimidine 97%
2-Amino-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD12923656
CAS Number: 93366-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR